missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citronellic acid, 94%
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Description
Citronellic acid was used in synthesis of stereoisomers of 3,11- dimethylnonacosan-2-one and 29-hydroxy-3,11-dimethylnonacosan-2-one.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Citronellic acid was used in synthesis of stereoisomers of 3,11- dimethylnonacosan-2-one and 29-hydroxy-3,11-dimethylnonacosan-2-one.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Citronellic acid was used in synthesis of stereoisomers of 3,11- dimethylnonacosan-2-one and 29-hydroxy-3,11-dimethylnonacosan-2-one.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Specifications
| Density | 0.923 |
| Boiling Point | 121°C to 122°C (1 mmHg) |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.4535 |
| Quantity | 25 g |
| UN Number | UN2810 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 170.25 |
| Percent Purity | 94% |
| Chemical Name or Material | Citronellic acid |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction