missing translation for 'onlineSavingsMsg'
Learn More

Citronellol, 95%

Product Code. 11439326
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439326 500 g 500g
11429326 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11439326 Supplier Thermo Scientific Alfa Aesar Supplier No. A19016.36
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 106-22-9 | C10H20O | 156.269 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-22-9
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
MDL Number MFCD00002935
InChI Key QMVPMAAFGQKVCJ-UHFFFAOYSA-N
Synonym citronellol, beta-citronellol, 3,7-dimethyl-6-octen-1-ol, dl-citronellol, cephrol, elenol, 6-octen-1-ol, 3,7-dimethyl, rodinol, 2,3-dihydrogeraniol, 2,6-dimethyl-2-octen-8-ol
PubChem CID 8842
ChEBI CHEBI:50462
IUPAC Name 3,7-dimethyloct-6-en-1-ol
SMILES CC(CCC=C(C)C)CCO

Specifications

Melting Point <-20°C
Density 0.858
Boiling Point 221°C to 222°C
Flash Point 98°C (208°F)
Odor Pleasant
Refractive Index 1.4575
Quantity 500 g
UN Number UN3082
Beilstein 1721507
Merck Index 14,2330
Formula Weight 156.27
Percent Purity 95%
Chemical Name or Material Citronellol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.