missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Citrus Clearing Solvent

Product Code. 10444720 Shop All Epredia Products
Click to view available options
Quantity:
4 x 3.8L (4 x 1 gal.)
Unit Size:
15.2 L
This item is not returnable. View return policy

Product Code. 10444720

Brand: Epredia™ 8301

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Replace xylene in both manual and automated system applications with Epredia™ Citrus Clearing Solvent.

  • D-limonene-based solvent has a slight citrus scent.
  • Higher flash point eliminates the need for storage in a flammable safety cabinet.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 4 x 3.8L (4 x 1 gal.)
compliance-icons
Product Identifier
  • Citrus Clearing Solvent
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.