missing translation for 'onlineSavingsMsg'
Learn More

CJ-023423 Hydrochloride Salt, Hydrate, TRC

Product Code. 30138397
Change view
Click to view available options
Quantity:
1 mg
25 mg
50 mg
Unit Size:
1mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138397 25 mg 25mg
30137665 50 mg 50mg
30005603 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138397 Supplier Toronto Research Chemicals Supplier No. TRCC53580825MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C26H29N5O3S• x(HCl), y(H2O)
Molecular Weight (g/mol) 491.6136461802
IUPAC Name 1-[2-[4-(2-ethyl-4,6-dimethylimidazo[4,5-c]pyridin-1-yl)phenyl]ethyl]-3-(4-methylphenyl)sulfonylurea
SMILES O=S(C1=CC=C(C)C=C1)(NC(NCCC2=CC=C(N3C(CC)=NC4=C3C=C(C)N=C4C)C=C2)=O)=O

Specifications

Quantity 25 mg
Synonym 2-Ethyl-4,6-dimethyl-1-[4-[2-[[[[(4-methylphenyl)sulfonyl]amino]carbonyl]amino]ethyl]phenyl]-1H-imidazo[4,5-c]pyridine Hydrochloride Salt Hydrate, AAT 007 Hydrochloride Salt Hydrate, CJ 023423 Hydrochloride Salt Hydrate, Grapiprant Hydrochloride Salt Hydrate, MR 10A7 Hydrochloride Salt Hydrate, N-[[[2-[4-(2-Ethyl-4,6-dimethyl-1H-imidazo[4,5-c]pyridin-1-yl)phenyl]ethyl]amino]carbonyl]-4-methylbenzenesulfonamide Hydrochloride Salt Hydrate, RQ 00000007 Hydrochloride Salt Hydrate, RQ 7 Hydrochloride Salt Hydrate, N-[[[2-[4-(2-Ethyl-4,6-dimethyl-1H-imidazo[4,5-c]pyridin-1-yl)phenyl]ethyl]amino]carbonyl]-4-methyl-benzenesulfonamide Hydrochloride Salt Hydrate
InChI Formula InChI=1S/C26H29N5O3S/c1-5-24-29-25-19(4)28-18(3)16-23(25)31(24)21-10-8-20(9-11-21)14-15-27-26(32)30-35(33,34)22-12-6-17(2)7-13-22/h6-13,16H,5,14-15H2,1-4H3,(H2,27,30,32)
Chemical Name or Material CJ-023423 Hydrochloride Salt, Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.