missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CL-XPosure™ Film (X-Ray Film)

Product Code. 12333850 Shop All Thermo Scientific Products
Change view
Click to view available options
Dimensions (LxW):
5 x 7 in. (12.5 x 17.5 cm)
8 x 10 in. (20 x 25 cm)
Unit Size:
50 sheets
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (LxW) unitSize
12333850 5 x 7 in. (12.5 x 17.5 cm) 50 sheets
10381544 8 x 10 in. (20 x 25 cm) 50 sheets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 12333850 Supplier Thermo Scientific™ Supplier No. 34088

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Capture results of ECL-based Western blots and other chemiluminescent protein or nucleic acid assays using this economically priced, clear-blue X-ray film.

Thermo Scientific™ CL-XPosure Film is an economically priced clear-blue X-ray film especially for detection and documentation of ECL-based Western blots and other chemiluminescent protein or nucleic acid assay methods.

CL-XPosure Film is an excellent phototgraphic film for use with enhanced chemiluminescence (ECL) substrates for horseradish peroxidase (HRP) or alkaline phosphatase (AP). Convenient sizes are offered for protein Western blotting, standard film cassettes and film developing equipment. The autoradiography film also works well for chemiluminescent or isotopic DNA and RNA methods, including Southern and Northern blotting and gel-shift assays (EMSA). The quality and price of this film make it the smart choice for life science research applications.

Highlights:

  • High performance – just as sensitive as other commercially available autoradiography films

  • Affordable – costs up to 75% less than other brands of comparable quality X-ray film

  • Convenient – available in five sheet sizes in packages containing 50 or 100 non-interleaved sheets

  • Perfect match – validated for use with Thermo Scientific SuperSignal Chemiluminescent Substrates and Thermo Scientific Pierce ECL Chemiluminescent Substrate

Ordering Information:

Available in 5 × 7in., 7 × 9.5in. , 9.5 × 11in., or 14 × 17in. sheets

Specifications

Formulation Blue-backed, silver-emulsion, two-sided autoradiography film
Sufficient For 50 uses
Quantity 50 Sheets
Dimensions (LxW) 5 x 7 in. (12.5 x 17.5 cm)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.