missing translation for 'onlineSavingsMsg'
Learn More

Hach™ CL17 Analyzer Reagent Solutions

Product Code. 13286790
Click to view available options
Type:
Free Chlorine Indicator Solution
Total Chlorine Buffer Solution
Format:
Solution
Analytes:
Free Chlorine
Total Chlorine
Unit Size:
473mL
Each
This item is not returnable. View return policy

Product Code. 13286790

Brand: Hach™ 2314011

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use with the CL17 Chlorine Analyzer (model 5440000) or bulk analytical methods to measure Total or Free Chlorine levels. Hach™ CL17 Analyzer Reagent Solutions include DPD Compound, Indicator Solution (for free or total chlorine) and Buffer Solution (for free or total chlorine).

The CL17 Chlorine Analyzer is a dependable online analyzer for colorimetric DPD analysis of free chlorine, total chlorine, or permanganate in disinfection applications. It provides accurate, reliable results and is EPA-approved for Free or Total Chlorine Analysis. Monthly preventative maintenance can be performed in 15 minutes or less with no special tools.

  • Select Free or Total Chlorine reagents (indicator and buffer solutions, depending on parameter being measured
  • The DPD compound is added to the bottle of Free or Total Chlorine Indicator Solution
  • The bottles of Indicator + DPD solution and Buffer solution (Free or Total) are loaded directly into the CL17 Chlorine Analyzer
  • The CL17 can operate unattended for up to 30 days, without daily maintenance

Specifications

Analytes Free Chlorine
Certifications/Compliance EPA
Format Solution
For Use With (Application) Cholorimetric Analysis of Free Chlorine
For Use With (Equipment) CL17 Chlorine Analyzer (man. no. 5440000)
No. of Tests 450
Quantity 473 mL
Range 0 - 5mg/L
Test Method DPD
Type Free Chlorine Indicator Solution
compliance-icons
Avvertenza
  • Danger
Categoria di pericolo
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1
Indicazioni di pericolo
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Consigli di prudenza
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato