missing translation for 'onlineSavingsMsg'
Learn More

Hach™ CL17 Analyzer Reagent Solutions

Product Code. 10050414
Click to view available options
Type:
Free Chlorine Indicator Solution
Total Chlorine Buffer Solution
Format:
Solution
Analytes:
Free Chlorine
Total Chlorine
Unit Size:
473mL
Each
This item is not returnable. View return policy

Product Code. 10050414

Brand: Hach™ 2263511

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use with the CL17 Chlorine Analyzer (model 5440000) or bulk analytical methods to measure Total or Free Chlorine levels. Hach™ CL17 Analyzer Reagent Solutions include DPD Compound, Indicator Solution (for free or total chlorine) and Buffer Solution (for free or total chlorine).

The CL17 Chlorine Analyzer is a dependable online analyzer for colorimetric DPD analysis of free chlorine, total chlorine, or permanganate in disinfection applications. It provides accurate, reliable results and is EPA-approved for Free or Total Chlorine Analysis. Monthly preventative maintenance can be performed in 15 minutes or less with no special tools.

  • Select Free or Total Chlorine reagents (indicator and buffer solutions, depending on parameter being measured
  • The DPD compound is added to the bottle of Free or Total Chlorine Indicator Solution
  • The bottles of Indicator + DPD solution and Buffer solution (Free or Total) are loaded directly into the CL17 Chlorine Analyzer
  • The CL17 can operate unattended for up to 30 days, without daily maintenance

Specifications

Analytes Total Chlorine
Certifications/Compliance EPA
Format Solution
For Use With (Application) Cholorimetric Analysis of Total Chlorine
For Use With (Equipment) CL17 Chlorine Analyzer (man. no. 5440000)
Quantity 473 mL
Range 0 - 5mg/L
Test Method DPD
Type Total Chlorine Buffer Solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.