Learn More
Phenomenex™ Claricep™ C18E SPE Columns, Telos Basic Alumina
Product Code. 31748945
Change view
Click to view available options
Bed Weight (Metric):
20 g
Unit Size:
Pack of 50
Product Code. Bed Weight (Metric) unitSize
31748945 20 g Pack of 50
1 options
Product Code. 31748945 Supplier Phenomenex™ Supplier No. 182003
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
Available in two flash media (irregular and spherical) with multiple chemistries, Claricep columns are optimized for resolution and high loading capacity. The deactivated silica surface promotes compound stability, excellent peak shape and high performance for both acidic and basic compounds.
  • Brand: Claricep
  • Technique: SP
  • Phase: Telos Basic Alumina
TRUSTED_SUSTAINABILITY
Specifications
Type Flash Column
Bed Weight (Metric) 20 g