Learn More
Phenomenex™ Claricep™ C8 Flash Column, Spherical Media, with 2 x Frit
Product Code. 31745497 Shop All Phenomenex Products
Change view
Click to view available options
Type:
Flash Column
Unit Size:
Each
Product Code. Type Unit Size
31745497 Flash Column Each
1 options
Product Code. 31745497 Supplier Phenomenex™ Supplier No. FC81401000
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Available in two flash media (irregular and spherical) with multiple chemistries, Claricep columns are optimized for resolution and high loading capacity. The deactivated silica surface promotes compound stability, excellent peak shape and high performance for both acidic and basic compounds.
  • Brand: Claricep
  • Technique: FLASH
  • Separation Mode: Reversed Phase
  • Phase: C8
  • Particle Type: Spherical
TRUSTED_SUSTAINABILITY
Specifications
Type Flash Column