Learn More
Phenomenex™ Claricep™ Flash Column Adaptor
Product Code. 31744473 Shop All Phenomenex Products
Change view
Click to view available options
Type:
Flash Column
Unit Size:
Each
Product Code. Type Unit Size
31744473 Flash Column Each
1 options
Product Code. 31744473 Supplier Phenomenex™ Supplier No. FHJT
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
Available in two flash media (irregular and spherical) with multiple chemistries, Claricep columns are optimized for resolution and high loading capacity. The deactivated silica surface promotes compound stability, excellent peak shape and high performance for both acidic and basic compounds.
  • Brand: Claricep
  • Technique: Accessory
TRUSTED_SUSTAINABILITY
Specifications
Type Flash Column