Learn More
Phenomenex™ Claricep™ LRC Filtration Column, 1/16 in. Sieve Plate
Product Code. 31748875 Shop All Phenomenex Products
Change view
Click to view available options
Type:
Flash Column
Unit Size:
Pack of 100
Product Code. Type Unit Size
31748875 Flash Column Pack of 100
1 options
Product Code. 31748875 Supplier Phenomenex™ Supplier No. FCL010
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Available in two flash media (irregular and spherical) with multiple chemistries, Claricep columns are optimized for resolution and high loading capacity. The deactivated silica surface promotes compound stability, excellent peak shape and high performance for both acidic and basic compounds.
  • Brand: Claricep
  • Technique: SP
  • Separation Mode: Multi-Mode
  • Phase: Mixed Mode
TRUSTED_SUSTAINABILITY
Specifications
Type Flash Column
Quantity 10 Pk.