Learn More
Phenomenex™ Claricep™ Neutral Alumina Sorbent for Flash Columns, Irregular Media, Particle Size 100 to 300 μm
Product Code. 31748900 Shop All Phenomenex Products
Change view
Click to view available options
Type:
Flash Column
Unit Size:
Each
Product Code. Type Unit Size
31748900 Flash Column Each
1 options
Product Code. 31748900 Supplier Phenomenex™ Supplier No. FCA130100N
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Available in two flash media (irregular and spherical) with multiple chemistries, Claricep columns are optimized for resolution and high loading capacity. The deactivated silica surface promotes compound stability, excellent peak shape and high performance for both acidic and basic compounds.
  • Brand: Claricep
  • Technique: FLASH
  • Phase: Alumina
  • Particle Type: Irregular
TRUSTED_SUSTAINABILITY
Specifications
Type Flash Column