Learn More
Phenomenex™ Claricep™ Screw-on Flash Colunm, Spherical
Product Code. 31743188 Shop All Phenomenex Products
Change view
Click to view available options
Bed Weight (Metric):
12 g
Unit Size:
Pack of 20
Product Code. Bed Weight (Metric) Unit Size
31743188 12 g Pack of 20
1 options
Product Code. 31743188 Supplier Phenomenex™ Supplier No. SNSP2300120
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Available in two flash media (irregular and spherical) with multiple chemistries, Claricep columns are optimized for resolution and high loading capacity. The deactivated silica surface promotes compound stability, excellent peak shape and high performance for both acidic and basic compounds.
  • Brand: Claricep
  • Technique: FLASH
  • Particle Type: Spherical
TRUSTED_SUSTAINABILITY
Specifications
Type Flash Column
Bed Weight (Metric) 12 g
Quantity 20 Pk.