missing translation for 'onlineSavingsMsg'
Learn More

Poulten Graf™ Class A Borosilicate Glass Measuring Cylinder

Product Code. 12368945
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
2000 mL
Graduations:
0.2 mL
0.5 mL
1 mL
20 mL
Unit Size:
Each
Pack of 2
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Graduations unitSize
12368945 50 mL 1 mL Pack of 2
12348945 10 mL 0.2 mL Pack of 2
12358945 25 mL 0.5 mL Pack of 2
12338945 2000 mL 20 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 12368945 Supplier Poulten Graf™ Supplier No. R264/WAC/E/2

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Base Type Hexagonal
Calibration To Contain
Scale Color Blue
Shape Round
Type Graduated Mixing Cylinder
Material Borosilicate Glass
Certifications/Compliance EN ISO 4788
Class A
Capacity (Metric) 50 mL
Includes Single Certificate, Stopper
Stopper Material Polypropylene
Graduations 1 mL
Autoclavable Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.