missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand Class II Biological Safety Cabinet

Product Code. 30564666 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Length (Metric) Exterior:
1.2 m
1.8 m
Plug Type:
UK
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Exterior Plug Type Unit Size
30564666 1.2 m UK Each
30564667 1.8 m UK Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30564666
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fisherbrand Class II Biological Safety Cabinet is an economical solution for high protection.

  • Fisherbrand Class II biological safety cabinet (BSC) provides both personal and product protection from biological hazards and contaminations.
  • This model is available in 2 sizes; 1.2 m and 1.8 m with 3 different plug and socket options (Type F, Type G, and Type E)
  • Equipped with technology to automate airflow compensation and alerts user to out of tolerance airflows that could compromise product and operator protections
  • H14 HEPA filters removes airborne contaminants
  • 10-degrees sloped front for exceptional comfort and ergonomics
  • SmartClean front window design to improve access to the work area for cleaning
  • Night Set Back Mode reduces airflow during low-usage periods
  • The BSC's comply with all applicable performance and safety requirements of EN12469:2000
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 12469
Chamber Material Stainless Steel (304)
Product Type Class II Biosafety Cabinet
Model Fisherbrand BSC 1.2
Noise Level <59 dBA
Outlets 2
Plug Type UK
Power Consumption 1440 W
Receptacles 2 Single, Rear Wall
Exhaust Air Filter HEPA H14 EN1822
Filter Efficiency 99.995% at MPPS
Includes UV light, armrest and adjustable floor stand
Energy Consumption (Reduced Flow Mode) 70 W
Lighting LED
Ports 2 x 2
Supply Air Filter HEPA H14 EN1822
Warranty 2 Years
Worksurface Area 120 x 45 cm
Dimensions (L x W x H) Exterior 130 x 80.2 x 152 cm
Length (Metric) Exterior 1.2 m
Depth (Metric) Interior 63 cm
Electrical Requirements 230 V, 50/60 Hz
Voltage 230 V
Energy Consumption (Operating Set Point) 200 W
Quantity 1 Ea.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.