Learn More
3M™ Clean-Trace™ ATP Water Test
A dedicated ATP water test designed to prevent sampling inaccuracy inherent in collecting a water sample with a swab.
Brand: 3M™ 7100074112
Additional Details : Weight : 0.20000kg
Description
As an integral part of the 3M™ Clean-Trace™ Hygiene Management System, the Clean-Trace Water Test uses adenosine tri-phosphate (ATP) to monitor the outcomes of manual cleaning of flexible endoscopes prior to sterilization or high-level disinfection.
- Simple to easy
- Powerful addition to your hygiene program
- Results measured in the 3M™ Clean-Trace™ NG Luminometer and provided in Relative Light Units (RLU) to allow rapid decisions on cleanliness status
Specifications
Water Testing for Surgical Instruments and Environmental Surfaces | |
100 Pk. | |
Hygiene Management System |