missing translation for 'onlineSavingsMsg'
Learn More

Phenomenex™ Cleanert™ Crimp Caps with Septa, 11 mm, No Slit

Product Code. 31745776 Shop All Phenomenex Products
Change view
Click to view available options
Septum Color:
Red/White
White/Red
Unit Size:
Each
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Septum Color Unit Size
31745776 Red/White Each
31744781 White/Red Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 31745776 Supplier Phenomenex™ Supplier No. C40114A
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Accessories for use with chromatography vials and in various chromatography applications.

  • Brand: Cleanert
  • Technique: Accessory
TRUSTED_SUSTAINABILITY

Specifications

Color Silver
Diameter (Metric) Outer 11 mm
Material Aluminum
Pre-slit No
Sterility Sterile
Septum PTFE/Silicone
Closure Size 11 mm
Marking Spot No
Autoclavable Autoclavable
Product Type Cap
Septum Color Red/White
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.