missing translation for 'onlineSavingsMsg'
Learn More

Phenomenex™ Cleanert™ Screw Caps with Septa, 13 mm, No Slit

Product Code. 31744888 Shop All Phenomenex Products
Change view
Click to view available options
Color:
Black
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
31744888 Black Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31744888 Supplier Phenomenex™ Supplier No. AV74100
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Accessories for use with chromatography vials and in various chromatography applications.

  • Brand: Cleanert
  • Technique: Accessory
TRUSTED_SUSTAINABILITY

Specifications

Color Black
Diameter (Metric) Outer 13 mm
Pre-slit No
Sterility Sterile
Septum PTFE/Silicone
Closure Size 13 mm
Marking Spot No
Autoclavable Autoclavable
Product Type Cap
Septum Color Red/White
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.