missing translation for 'onlineSavingsMsg'
Learn More

Phenomenex™ Cleanert™ Screw Caps with Septa, 22 mm, No Slit

Product Code. 31748344 Shop All Phenomenex Products
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
31748344 White Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31748344 Supplier Phenomenex™ Supplier No. AV79100
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Accessories for use with chromatography vials and in various chromatography applications.

  • Brand: Phenomenex
  • Technique: Accessory
TRUSTED_SUSTAINABILITY

Specifications

Color White
Diameter (Metric) Outer 22 mm
Pre-slit No
Sterility Sterile
Septum PTFE/Silicone
Closure Size 22 mm
Marking Spot No
Autoclavable Autoclavable
Product Type Cap
Septum Color Natural
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.