Learn More
Tork Cleaning Cloth W1/2/3
GREENER_CHOICE
Product Code. 17268730 Shop All Tork Products
Click to view available options
Color:
Blue
Unit Size:
Each
Product Code. 17268730
Brand: Tork 510237
Only null left
Add to basket
Add to basket
The multipurpose Tork Cleaning Cloth is soft and flexible, yet strong. It is ideal for most lighter cleaning tasks, and quickly absorbs both oils and liquids, which makes your cleaning efforts easier.
  • Tackle a range of tasks with our Tork Cleaning Cloth - it's suitable with most solvents, disinfectants and detergents.
  • Replace unsightly rags and rental towels with these durable cloths for repeated use.
  • Swiftly remove oil, grease, lubricants, and dirt, thanks to their high absorbency.
  • Effortlessly complete lighter cleaning tasks with its soft and flexible feel.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleaning Cloth
Length (English) 166.23 yd.
Length (Metric) 152 m
Width (English) 12.4 in.
Width (Metric) 31.5 cm
Autoclavable Not Autoclavable
Certifications/Compliance FSC MIX Credit, ISO 9001, ISO 14001
Color Blue
Disposable Limited-use
Packaging Quantity 1 Roll
Ply 1-ply
Certifications/Compliance (Europe) FSC MIX Credit, ISO 9001, ISO 14001