missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Cleaning Detergent rea-clean™

Product Code. 10160661
Change view
Click to view available options
Volume (Metric):
1 L
30 L
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10160661 1 L Each
10479322 30 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10160661 Supplier Hirschmann™ Supplier No. 9790101
  • ONLINE EXCLUSIVE PRICE
    25.35 €
    22.15 € / Pack of 1
    Save 3.20 €
    12% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Bio degradable cleaning detergent for rapid, automatic and careful cleaning of materials and surfaces especially made of plastics.

  • Biodegradable cleaner
  • Surface cleaning
  • Phosphate-free and self-neutralising

Specifications

Product Type Phosphate-free Cleaning Concentrate
Form Liquid
Product Line rea-clean
Description Biodegradable, fat dissolving, gentle on materials
For Use With (Equipment) All sufaces in labs
Quantity 1 Refill Bottle
Volume (Metric) 1 L
compliance-icons
Product Identifier
  • rea-clean Bio-Reiniger
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.