Learn More
BD Accuri™ Cleaning Solution Concentrate
Used for cleaning the components in flow cytometers, including the Accuri™ C6. BD™ Accuri™ Cleaning Solution Concentrate makes 1L cleaning solution that complies with RUO regulatory status.
Brand: BD 653157
Code : NEW
Additional Details : CAS Number : 77-92-9 Weight : 1.12000kg
Description
To ensure consistent instrument functioning, always clean and rinse the instrument before turning it off at the end of the day and immediately after running viscous samples or dyes.
- Application: Flow cytometry
- Added to 1L deionized water to make cleaning solution for the BD Accuri™ C6 Flow Cytometer
- Lot number and expiration date listed on label of each vial
- For research only (RUO)—Not for therapeutic or diagnostic use
Physical properties:
- pH at 20°C: 2.5
- Boiling point: 130°C
- Density: 1.12g/cm3 at 20°C
- Soluble in water
Specifications
Citric acid 25%, Glycollic acid 10%, Water 55% | |
Liquid | |
Yellowish |
Concentrated cleaning solution for flow cytometers | |
RUO | |
1 L |
Safety and Handling
- Cleaning Concentrate Solution
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : citric acid, glycollic acid