Learn More
Cole-Parmer Cleanroom Female Luer to Hose Barb Fittings, Straight Adapter, CrystalVu™
Product Code. 30053404 Shop All Cole-Parmer Products
Change view
Click to view available options
Diameter (English) Inner:
1/16 in.
1/4 in.
1/8 in.
3/16 in.
3/32 in.
5/32 in.
Unit Size:
Pack of 10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Inner unitSize
30053404 3/32 in. Pack of 10
30053071 3/16 in. Pack of 10
30054213 1/4 in. Pack of 10
30053954 5/32 in. Pack of 10
30053755 1/16 in. Pack of 10
30053380 1/8 in. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 30053404 Supplier Cole-Parmer Supplier No. 5010966
  • ONLINE EXCLUSIVE PRICE
    9.15 €
    7.57 € / Pack of 10
    Save 1.58 €
    17% Off
Estimated Shipment: 12-10-2026
Add to basket
Add to basket
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Forms a leak-free luer connection with a quick simple turn. Single barb design tightly grips the tubing. USP Class VI, EU 2002/72/EC, and RoHS compliant
Specifications
Certifications/Compliance Non-DEHP;Non-Phthalate;RoHS;USP
Diameter (English) Inner 3/32 in.
Packaging Quantity 10/Pk.
Inlet Female Luer x Hose Barb