Learn More
Cole-Parmer Cleanroom Hose Barb Fittings, Straight Union, Natural Kynar™
Product Code. 30053518 Shop All Cole-Parmer Products
Change view
Click to view available options
Diameter (English) Inner:
1 in.
1/16 in.
1/2 in.
1/4 in.
1/8 in.
3/16 in.
3/32 in.
3/4 in.
3/8 in.
5/16 in.
5/32 in.
5/8 in.
Unit Size:
Pack of 10
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Inner unitSize
30053518 5/16 in. Pack of 10
30053310 5/32 in. Pack of 10
30054295 3/16 in. Pack of 10
30053668 1/8 in. Pack of 10
30053563 1/16 in. Pack of 10
30053751 5/8 in. Pack of 10
30054274 3/4 in. Pack of 10
30053662 1/2 in. Pack of 10
30054149 1/4 in. Pack of 10
30053840 3/8 in. Pack of 10
30053482 1 in. Pack of 10
30053195 3/32 in. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 30053518 Supplier Cole-Parmer Supplier No. 5010837
Only null left
Add to basket
Add to basket
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Quick and easy assembly without tools. Single barb design tightly grips the tubing. Biopharma compatible and USP 661 and BPOG compliant
Specifications
Certifications/Compliance USP;Non-DEHP;Non-Phthalate
Diameter (English) Inner 5/16 in.
Packaging Quantity 10/Pk.
Outlet Port Hose Barb Union
Sterilization Method E-beam/gamma irradiation, EtO, or autoclave up to 135°C
Shape Straight
Color Natural