Learn More
Cole-Parmer Cleanroom Port Disk Hose Barb Fittings, Flat Plate, Natural Kynar™ Flex
Product Code. 30054341 Shop All Cole-Parmer Products
Click to view available options
Diameter (English) Inner:
1 in.
1/2 in.
1/4 in.
1/8 in.
3/16 in.
3/4 in.
3/8 in.
5/16 in.
5/32 in.
5/8 in.
Unit Size:
Pack of 10
Product Code. 30054341
Brand: Cole-Parmer 5011713
Only null left
Add to basket
Add to basket
Weld to single-use bags to transfer cell culture, drug compounds, and other biological fluids
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Flexible, smooth flat base for easy attachment. Ultrasonic, radio frequency, or thermal weld to Kynar bags. Meets FDA requirements 21 CFR 177.1520(c), 21 CFR 176.170(c), 21 CFR 177.2600(c)(4)(i), NSF/ANSI Standard 51 and 61, and RoHS compliant
Specifications
Certifications/Compliance FDA;Non-DEHP;Non-Phthalate;RoHS
Diameter (English) Inner 3/4 in.
Packaging Quantity 10/Pk.
Outlet Port Port Disk x Hose Barb
Sterilization Method 121°C
Shape Straight
Color Natural