Learn More
Cole-Parmer Cleanroom Rotating Male Luer Lock to Barb Fittings, Straight Adapter, Animal-Derivative-Free Polypropylene
Product Code. 30054007 Shop All Cole-Parmer Products
Change view
Click to view available options
Diameter (English) Inner:
1/16 in.
1/4 in.
1/8 in.
3/16 in.
3/32 in.
5/32 in.
Unit Size:
Pack of 10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Inner Unit Size
30054007 3/32 in. Pack of 10
30053987 3/16 in. Pack of 10
30053583 1/16 in. Pack of 10
30053530 5/32 in. Pack of 10
30053648 1/4 in. Pack of 10
30053692 1/8 in. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 30054007 Supplier Cole-Parmer Supplier No. 5011454
  • ONLINE EXCLUSIVE PRICE
    5.10 €
    4.25 € / Pack of 10
    Save 0.85 €
    16% Off
Estimated Shipment: 03-11-2026
Add to basket
Add to basket
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Connect to a female luer body to form a leak-free fluid path. Single barb design tightly grips the tubing. Meets FDA requirements 21 CFR 177.1520(a)(3)(i) and (c)3.1a and RoHS compliant
Specifications
Certifications/Compliance ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS
Diameter (English) Inner 3/32 in.
Material Polypropylene
Packaging Quantity 10/Pk.
Inlet Male Luer x Hose Barb