missing translation for 'onlineSavingsMsg'
Learn More

PURUS™ Cleanroom Shoes

Product Code. 31789575 Shop All Purus Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Unit Size:
6 pairs
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
31789575 2X-Large 6 pairs
31789574 X-Small 6 pairs
31789572 Small 6 pairs
31789571 Medium 6 pairs
31789570 Large 6 pairs
31789573 X-Large 6 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 31789575 Supplier PURUS™ Supplier No. CRSHWXXL
  • ONLINE EXCLUSIVE PRICE
    670.75 €
    536.77 € / 6 pairs
    Save 133.98 €
    19% Off
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

PURUS™ Cleanroom Shoes provide comfort and durability when working in cleanroom environments.

  • Smooth surface for day-long comfort
  • Antistatic
  • Autoclavable
  • Suitable for use in ISO 4 to 6 (Class 10 to Class 1000) cleanroom
  • Non-slip sole
  • Special conical design prevents spilled liquids from entering clog
  • Heel strap
  • Anatomic Heel Basin
  • Toe Grabber

Specifications

Certifications/Compliance ASTM F2913
Color White
For Use With (Application) Pharmaceutical, Biotech, Semiconductor, Cleanrooms, Hospitals
Material Thermoplastic Elastomer
Size 2X-Large
Toe Style Plain Toe
Packaging Quantity 6/Cs.
Product Type Cleanroom Shoes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.