Learn More
Micronova™ Cleanser
Product Code. 13117296 Shop All Micronova Products
Product Code. 13117296 Supplier Micronova™ Supplier No. IC120
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Micronova LoNa™ hand soap and glove cleanser is a low sudding soap for cleanroom use.
LoNa, low sodium, controlled environment, low sudsing, pH balanced for smooth skin, amber
Specifications
Description LoNa™ hand soap and glove cleanser
For Use With (Application) Fomulated for high-tech cleanrooms before and after gowning
Packaging Pump bottle
Volume (Metric) 500 mL