missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diversey™ ClearKlens Cleanisald Disinfectant
Description
- Blend of quaternary ammonium and alkyl amines which dilutes to 5L for cleaning and disinfection of hard surfaces
- For mopping and wiping applications
- Supplied with certificates of analysis and sterility
- Broad spectrum of activity
Specifications
Specifications
| pH | 10 (neat) |
| Sterility | Sterile |
| Filtered | 0.2 μm |
| Quantity | 4/Cs. |
| Volume (Metric) | 5 L |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?