missing translation for 'onlineSavingsMsg'
Learn More

Diversey™ ClearKlens DE Disinfectant

Product Code. 15390308
Change view
Click to view available options
Quantity:
4/Cs.
6/Cs.
Unit Size:
20 L
5.4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15390308 6/Cs. 5.4 L
15380308 4/Cs. 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15390308 Supplier Diversey™ Supplier No. 6069643

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Disinfect surfaces in industrial and cleanroom applications. Diversey ClearKlens DE Disinfectant is based on 70% denaturated ethanol.

  • 70% ethanol, 30% water for injection disinfectant
  • For use on contact surfaces, gloved hands, instruments, and utensils
  • Gamma irradiated (900mL) or sterile filled (5L) and double bagged
  • Detex labelled for proof of irradiation (900mL)
  • Supplied with certificates of analysis, irradiation, endotoxin, and sterility where applicable
  • 900mL trigger spray incorporates anti-suck technology

Specifications

pH ∼7
Sterility Sterile
Composition 70% ethanol, 30% water
Content And Storage Store at Room Temperature
Filtered Filtered
Includes Trigger spray
Quantity 6/Cs.
Volume (Metric) 5.4 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.