missing translation for 'onlineSavingsMsg'
Learn More

Diversey™ ClearKlens IPA VH1 Disinfectant
BIOPHARMA

Product Code. 17683095
Change view
Click to view available options
Volume (Metric):
500 mL
1 L
5 L
Description:
For AT, BE, CH, DE, FR, GB, IE, IT, NL only
Unit Size:
10 L
12 L
6 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Description Unit Size
17683095 5 L For AT, BE, CH, DE, FR, GB, IE, IT, NL only 10 L
17693095 500 mL For AT, BE, CH, DE, FR, GB, IE, IT, NL only 6 L
17603105 1 L For AT, BE, CH, DE, FR, GB, IE, IT, NL only 12 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17683095 Supplier Diversey™ Supplier No. 101105065
  • ONLINE EXCLUSIVE PRICE
    132.00 €
    112.84 € / 10 L
    Save 19.16 €
    14% Off
 Restricted Item
Estimated Shipment: 15-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ready to use disinfectant for non-porous hard surfaces and gloves

ClearKlens IPA is a non-sterile, ready to use disinfectant for the Life Sciences industry. It is used as general purpose mid-shift and terminal disinfectant. ClearKlens IPA is based on 70% isopropyl alcohol. ClearKlens IPA is available as 500 mL trigger spray, and as 1 L bottle or 5 L canister.

Key properties

  • ClearKlens IPA is a fast acting disinfectant spray for use in mid-shift and terminal disinfection applications.
  • It has a broad spectrum of efficacy against most vegetative forms of micro- organisms.
  • ClearKlens IPA is safe for use on food contact surfaces and evaporates rapidly, leaving no residue. It is suitable for the disinfection of alcohol-safe surfaces, machines and components (conveyors, fillers, bottle inspectors, water-sensitive components, etc.)
  • BPR approved pack and formulation

Benefits

  • Filtered at 0.2 μm
  • Ready to use
  • Effective broad-spectrum activity
  • Non foaming
  • Suitable for use in moisture sensitive areas
  • Readily evaporates leaving no residues

Specifications

Product Type Surface Disinfectant
pH 6 to 9
Sterility Non-sterile
Composition 70% IPA/30% deionized water
Container Type Canister
Description For AT, BE, CH, DE, FR, GB, IE, IT, NL only
Quantity 2 Pk.
Volume (Metric) 5 L
compliance-icons compliance-icons
Product Identifier
  • ClearKlens IPA VH1
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P370+P378-In case of fire: Use alcohol-resistant foam to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: propan-2-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.