missing translation for 'onlineSavingsMsg'
Learn More

Diversey™ ClearKlens Plus VH5 S Liquid Detergent

Product Code. 17986110
Change view
Click to view available options
Volume (Metric):
30 mL
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
17986110 30 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17986110 Supplier Diversey™ Supplier No. 101109135
  • ONLINE EXCLUSIVE PRICE
    1420.00 €
    1214.85 € / Pack of 100
    Save 205.15 €
    14% Off
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ClearKlens Plus is a sterile, concentrated, neutral, high-foaming liquid detergent for the life sciences industries. It is used for the manual cleaning of process equipment, floors and walls in cleanrooms.

  • ClearKlens Plus contains a concentrated blend of high foaming detergents and emulsifiers
  • Balanced neutral formulation is particularly effective against product residues found in the pharmaceutical and cosmetic industries
  • Can be used on most types of surfaces, including plastics and soft metals such as aluminum
  • Packaged in 30 mL double bagged unit doses, which have been irradiated at 25 kGy minimum for secure transfer into cleanroom
  • Certificate of analysis (COA) for each batch available

Specifications

Product Type Liquid Detergent
Color-coded No
Form Concentrated Liquid
pH Neutral
Product Line ClearKlens
Scent Unscented
Sterility Sterile
Autoclavable Not Autoclavable
Color Clear
Disposable Single-use
Filtered Non-filtered
Flammable No
Quantity 100 Bottles
Volume (Metric) 30 mL
Show More Show Less
compliance-icons
Product Identifier
  • ClearKlens Plus VH5
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Supplemental information
  • MIXTURE LIST-Contain: sodium alkylbenzenesulphonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.