missing translation for 'onlineSavingsMsg'
Learn More

Diversey™ ClearKlens Release VH45 Detergent
BIOPHARMA

Product Code. 17673245
Change view
Click to view available options
Volume (Metric):
20 L
Unit Size:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
17673245 20 L 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17673245 Supplier Diversey™ Supplier No. 7520002
  • ONLINE EXCLUSIVE PRICE
    188.00 €
    146.18 € / 20 L
    Save 41.82 €
    22% Off
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ClearKlens Release is a low foaming, mixed acid liquid for use in a wide range of applications in the pharmaceutical, cosmetic industries, and research centers.

ClearKlens Release can also be used as a descaler for CIP of cold and heat treated surfaces.

Key properties

  • ClearKlens Release is very efficient for tablet coating removal in the pharma industry.
  • ClearKlens Release meets the customer/application requirement of low phosphorus (min. 20 g/kg P) which improves the cleaning performance.
  • ClearKlens Release is low foaming above 40°C and is suitable for use in tunnel washer applications under conditions of high pressure and turbulence.
  • ClearKlens Release can be used as a low foam detergent descaler in including in research centers CIP applications.
  • ClearKlens Release is a conductive liquid and is suitable for automatic dosing and control.

Benefits

  • Low foaming properties above 40°C in tunnel washer applications under conditions of high pressure and turbulence, provides easy rinsing and improved cleaning performance.
  • Low level of phosphorus.
  • Suitable for automatic dosing and control by conductivity, ensuring consistent delivery of product.

Specifications

Form Liquid
pH 2.2
Sterility Non-sterile
Composition Acidic detergent descaler
Description 1% solution at 20°C
Volume (Metric) 20 L
compliance-icons
Product Identifier
  • ClearKlens Release VH45
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Citric acid, phosphoric acid, methanesulphonic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.