missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ CLED Agar
Isolate, enumerate and presumptively identify all pathogens occurring in urinary tract infections with this traditional media which provides good colonial differentiation.
14.00 €
Specifications
Description | CLED Agar |
---|---|
Format | 90mm Monoplate |
Product Type | Agar |
Packaging Type | Flow Wrapped - Single Layer |
Quantity | 10/Pk. |
Product Code | Brand | Description | Quantity | Manufacturing Origin | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Manufacturing Origin | Price | Quantity & Availability | |||
12992568
|
Thermo Scientific™
PO0120A |
CLED Agar | 10/Pk. | United Kingdom |
14.00 €
Pack of 10 |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||
Description
Detect common urinary tract pathogens using Thermo Scientific™ CLED Agar, a traditional media which provides good colonial differentiation, clear diagnostic characteristics and prevents swarming from Proteus spp. due to the electrolyte deficiency in the formulation.
Isolate, enumerate and presumptively identify all UTI related microorganisms with this traditional urinary bacteriology medium which provides a valuable non-inhibitory diagnostic for plate culture.This product may not be available in all countries or territories. Please consult your local representative for availability.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
CLED Agar | |
Agar | |
10/Pk. |
90mm Monoplate | |
Flow Wrapped - Single Layer |