missing translation for 'onlineSavingsMsg'
Learn More

Clenhexerol, TRC

Product Code. 30134272
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134272 100 mg 100mg
30005170 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134272 Supplier Toronto Research Chemicals Supplier No. TRCC572600100MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38339-23-0
Molecular Formula C14H22Cl2N2O
Molecular Weight (g/mol) 305.24
Synonym 4-Amino-3,5-dichloro-α-[[(1,1-dimethylbutyl)amino]methyl]benzenemethanol;
IUPAC Name 1-(4-amino-3,5-dichlorophenyl)-2-(2-methylpentan-2-ylamino)ethanol
SMILES ClC1=C(N)C(Cl)=CC(C(O)CNC(C)(C)CCC)=C1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C14H22Cl2N2O/c1-4-5-14(2,3)18-8-12(19)9-6-10(15)13(17)11(16)7-9/h6-7,12,18-19H,4-5,8,17H2,1-3H3
Formula Weight 304.1109
Chemical Name or Material Clenhexerol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.