missing translation for 'onlineSavingsMsg'
Learn More

Clenpenterol Hydrochloride, TRC

Product Code. 30137568
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137568 100 mg 100mg
30007517 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137568 Supplier Toronto Research Chemicals Supplier No. TRCC572800100MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37158-47-7
Molecular Formula C13 H20 Cl2 N2 O . Cl H
Molecular Weight (g/mol) 327.68
Synonym Benzenemethanol, 4-amino-3,5-dichloro-α-[[(1,1-dimethylpropyl)amino]methyl]-, hydrochloride (1:1), Benzenemethanol, 4-amino-3,5-dichloro-α-[[(1,1-dimethylpropyl)amino]methyl]-, monohydrochloride (9CI), 4-Amino-3,5-dichloro-α-[[(1,1-dimethylpropyl)amino]methyl]benzenemethanol hydrochloride (1:1), Clenpenterol hydrochloride
IUPAC Name 1-(4-amino-3,5-dichlorophenyl)-2-(2-methylbutan-2-ylamino)ethanol;hydrochloride
SMILES Cl.CCC(C)(C)NCC(O)c1cc(Cl)c(N)c(Cl)c1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C13H20Cl2N2O.ClH/c1-4-13(2,3)17-7-11(18)8-5-9(14)12(16)10(15)6-8;/h5-6,11,17-18H,4,7,16H2,1-3H3;1H
Formula Weight 326.0719
Percent Purity >95%
Chemical Name or Material Clenpenterol hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.