missing translation for 'onlineSavingsMsg'
Learn More

Clevudine, TRC

Product Code. 30358778
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30358778 100 mg 100mg
30354729 10 mg 10mg
30349324 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30358778 Supplier Toronto Research Chemicals Supplier No. TRCC574150100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 163252-36-6
Molecular Formula C10H13FN2O5
Molecular Weight (g/mol) 260.22
Synonym 1-(2'-Deoxy-2'-fluoro-beta-L-arabinofuranosyl)-5-methyluracil, L-FMAU, 1-(2-Deoxy-2-fluoro-beta-L-arabinofuranosyl)-5-methyl-2,4(1H,3H)-pyrimidinedione
IUPAC Name 1-[(2S,3R,4S,5S)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione
SMILES CC1=CN([C@H]2O[C@@H](CO)[C@H](O)[C@H]2F)C(=O)NC1=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H13FN2O5/c1-4-2-13(10(17)12-8(4)16)9-6(11)7(15)5(3-14)18-9/h2,5-7,9,14-15H,3H2,1H3,(H,12,16,17)/t5-,6+,7-,9-/m0/s1
Formula Weight 260.0808
Chemical Name or Material Clevudine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.