missing translation for 'onlineSavingsMsg'
Learn More

Nickel Electro™ Clifton™ Stainless Steel Electric Circular Boiling Bath

Product Code. 12728868
Change view
Click to view available options
Capacity (Metric):
3.24 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
12728868 3.24 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12728868 Supplier Nickel Electro™ Supplier No. 1026E
  • ONLINE EXCLUSIVE PRICE
    451.00 €
    390.36 € / Pack of 1
    Save 60.64 €
    13% Off
View all promotions
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Nickel Electro™ Clifton™ Stainless Steel Electric Circular Boiling Bath maintains a fixed temperature and is fitted with a concentric ringed lid to vary the hole size.

  • The steam produced provides an effective source of heat for samples that may be otherwise thermally damaged by a naked flame
  • Used for applications to distill/concentrate liquids, melt agar and other solids
  • Metallurgical analysis, protein analysis and bacteriological examinations
  • Features an integral constant level device to maintain constant water level during boiling and supplied with a set of rings to vary the diameter of aperture to accommodate different sizes of vessels of evaporating dishes and glassware, including round-bottom flasks.
  • Fitted with a sheathed immersion element and a one shot thermal cut out for safety.
  • Always connect the constant level with laboratory tubing to a water supply, adjust the flow to gently top up the water level, whilst maintaining boiling.
  • The Constant water level has a visible water spout for checking water is topping up the Boiling bath in use and avoids the chances of boiling dry.
  • Manual reset cut out
  • Warranty: 2 year

Specifications

Description Stainless steel boiling bath
Capacity (Metric) 3.24 L
Type Electric Circular Boiling Bath
Heater Settings (Low, Medium, High) Constant boiling no control
Material Stainless Steel
Temperature (Metric) Maximum 100°C
Warranty 2 year
Height (Metric) 200 mm
Diameter (Metric) 203 mm
Diameter (Metric) Min. Opening 42 mm
Shipping Weight (Metric) 2 kg
Voltage 230 V
Wattage 1000 W
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.