missing translation for 'onlineSavingsMsg'
Learn More

Nickel Electro™ Clifton™ Stainless Steel Rectangular Electric Boiling Bath, 6 x 80mm Holes

Product Code. 12778868
Change view
Click to view available options
Electrical Requirements:
230 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Electrical Requirements Unit Size
12778868 230 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12778868 Supplier Nickel Electro™ Supplier No. 1101E
  • ONLINE EXCLUSIVE PRICE
    1002.64 €
    687.76 € / Pack of 1
    Save 314.88 €
    31% Off
View all promotions
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rectangular, stainless steel electrically heated boiling bath, with fixed temperature for boiling water, fitted with concentric ringed lids to vary the hole size.

  • The steam produced provides an effective source of heat for samples:
    • that may be otherwise thermally damaged by a naked flame
    • used for applications to distil/concentrate liquids, melt agar and other solids
    • metallurgical analysis, protein analysis and bacteriological examinations
  • Features an integral constant level device to maintain constant water level during boiling and supplied with sets of rings to vary the diameter of each hole to accommodate different sizes of vessels of evaporating dishes and glassware, including round-bottom flasks simultaneously.
  • Fitted with a sheathed immersion element and a one shot thermal cut out for safety.
  • The bath provides constant boiling water for gentle heating of samples and in steam.
  • Accessory ER-1 Energy Regulator can be connected to boiling bath allowing control of the amount of boiling from a gentle simmer to a vigorous boil.
  • Always site the Boiling Bath on a heat proof surface.
  • Warranty: 2 year

Safety Features:

  • Always connect the constant level with laboratory tubing to a water supply, adjust the flow to gently top up the water level, whilst maintaining boiling.
  • The Constant water level has a visible water spout for checking water is topping up the Boiling bath in use and avoids the chances of boiling dry.
  • Manual reset cut out.
  • Always read the instruction book

Specifications

Description Stainless steel rectangular 6 hole boiling bath with concentric rings
Type Stainless Steel Rectangular 6 Hole Boiling Bath with Concentric Rings
Includes Concentric rings
Material Stainless Steel
Min. Opening Diameter (All Rings Removed) 80 mm
No. of Positions 6
Temperature (Metric) Maximum 100°C
Warranty 2 Years
Dimensions (L x W x H) 321 x 219 x 90 mm
Width (Metric) 321 mm
Diameter (Metric) 30 mm
Shipping Weight (Metric) 2.7 kg
Electrical Requirements 230 V
Voltage 230 V
Wattage 1000 W
Frequency 50/60 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.