missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Globe Scientific Clinical Chemistry Analyzer Sample Cups
Specifications
Specifications
| Closure Type | Snap |
| For Use With (Application) | Designed for use on automated instrumentation |
| For Use With (Equipment) | Roche cobas Mira™ analyzer |
| Color | Yellow |
| Material | Polyethylene |
| Product Type | Sample Cup |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Ser du en mulighed for forbedring?