Learn More
Sherwood Scientific™ Clinical Flame Photometer
Product Code. 12991801
Change view
Click to view available options
Type:
2-Channel Clinical Flame Photometer
Unit Size:
Each
Product Code. Type Unit Size
12991801 2-Channel Clinical Flame Photometer Each
1 options
Product Code. 12991801 Supplier Sherwood Scientific™ Supplier No. 47542000
  • ONLINE EXCLUSIVE PRICE
    10685.00 €
    9356.78 € / Pack of 1
    Save 1328.22 €
    12% Off
 Restricted Item
Estimated Shipment: 02-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Clinical single / dual element displayed 240V 50Hz Na with K, Na with Li 390mm x 345mm x 510mm Sherwood Scientific
Specifications
Gases Measured Na with K, Na with Li
Type 2-Channel Clinical Flame Photometer