missing translation for 'onlineSavingsMsg'
Learn More

Clobetasol 17-Propionate, TRC

Product Code. 30135239
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135239 1 g 1g
30134320 50 mg 50mg
18002540 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135239 Supplier Toronto Research Chemicals Supplier No. TRCC5835001G
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25122-46-7
Molecular Formula C25 H32 Cl F O5
Molecular Weight (g/mol) 466.97
Synonym Pregna-1,4-diene-3,20-dione, 21-chloro-9-fluoro-11β,17-dihydroxy-16β-methyl-, 17-propionate (8CI), (11β,16β)-21-Chloro-9-fluoro-11-hydroxy-16-methyl-17-(1-oxopropoxy)pregna-1,4-diene-3,20-dione, 21-Chloro-21-deoxybetamethasone 17-propionate, CCl 4725, CGP 9555, Clobederm, Clobesol, Clobetasol 17-propionate, Clobetasol propionate, Clobex, Dermoval, Dermovate, Dermoxin, Dermoxinale, GR 2/925, Karison, Olux, Olux-E, Skin Cap, Temovate, Tenovate
IUPAC Name [(8S,9R,10S,11S,13S,14S,16S,17R)-17-(2-chloroacetyl)-9-fluoro-11-hydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] propanoate
SMILES CCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)CCl

Specifications

Quantity 1 g
InChI Formula InChI=1S/C25H32ClFO5/c1-5-21(31)32-25(20(30)13-26)14(2)10-18-17-7-6-15-11-16(28)8-9-22(15,3)24(17,27)19(29)12-23(18,25)4/h8-9,11,14,17-19,29H,5-7,10,12-13H2,1-4H3/t14-,17-,18-,19-,22-,23-,24-,25-/m0/s1
Formula Weight 466.1922
Percent Purity >95%
Chemical Name or Material Clobetasol Propionate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.