missing translation for 'onlineSavingsMsg'
Learn More

Clobetasol propionate, MedChemExpress

Product Code. 16426246
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16426246 100 mg 100mg
16491086 200 mg 200mg
16471676 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16426246 Supplier Medchem Express Supplier No. HY13600
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Clobetasol propionate is a potent and selective CYP3A5 inhibitor with an IC50 of 0.206 μM. Clobetasol propionate has no inhibiting on CYP3A4 or other major CYPs. Clobetasol propionate is a corticosteroid and has the potential for psoriasis and other dermatoses research.

Clobetasol propionate is a potent and selective CYP3A5 inhibitor with an IC50 of 0.206 μM. Clobetasol propionate has no inhibiting on CYP3A4 or other major CYPs. Clobetasol propionate is a corticosteroid and has the potential for psoriasis and other dermatoses research.

Specifications

Chemical Name or Material Clobetasol propionate
CAS 25122-46-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C25H32ClFO5
Quantity 100 mg
Solubility Information DMSO : ≥ 100 mg/mL (214.15 mM)
SMILES C[C@@]12[C@](C(CCl)=O)(OC(CC)=O)[C@@H](C)C[C@@]1([H])[C@]3([H])CCC4=CC(C=C[C@]4(C)[C@@]3(F)[C@@H](O)C2)=O
Molecular Weight (g/mol) 466.97
Formula Weight 466.97
Percent Purity 99.85%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.