missing translation for 'onlineSavingsMsg'
Learn More

Clobetasone 17-Butyrate, TRC

Product Code. 30136578
Change view
Click to view available options
Quantity:
25 mg
100 mg
1 g
Unit Size:
100mg
1g
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136578 1 g 1g
18080440 100 mg 100mg
18050840 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136578 Supplier Toronto Research Chemicals Supplier No. TRCC5835501G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25122-57-0
Molecular Formula C26 H32 Cl F O5
Molecular Weight (g/mol) 478.98
Synonym Pregna-1,4-diene-3,11,20-trione, 21-chloro-9-fluoro-17-hydroxy-16β-methyl-, butyrate (8CI), (16β)-21-Chloro-9-fluoro-16-methyl-17-(1-oxobutoxy)pregna-1,4-diene-3,11,20-trione, Butyric acid, ester with 21-chloro-9-fluoro-17-hydroxy-16β-methylpregna-1,4-diene-3,11,20-trione (8CI), CCl 5537, Clobetasone 17-butyrate, Clobetasone butyrate, Emovate, Eumovate, GR 2/1214, Kindavate, Molivate, SN 203
IUPAC Name [(8S,9R,10S,13S,14S,16S,17R)-17-(2-chloroacetyl)-9-fluoro-10,13,16-trimethyl-3,11-dioxo-7,8,12,14,15,16-hexahydro-6H-cyclopenta[a]phenanthren-17-yl] butanoate
SMILES CCCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)C(=O)C[C@]12C)C(=O)CCl

Specifications

Quantity 1 g
InChI Formula InChI=1S/C26H32ClFO5/c1-5-6-22(32)33-26(21(31)14-27)15(2)11-19-18-8-7-16-12-17(29)9-10-23(16,3)25(18,28)20(30)13-24(19,26)4/h9-10,12,15,18-19H,5-8,11,13-14H2,1-4H3/t15-,18-,19-,23-,24-,25-,26-/m0/s1
Formula Weight 478.1922
Chemical Name or Material Clobetasone Butyrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.