missing translation for 'onlineSavingsMsg'
Learn More

Clobetasone 17-Propionate, TRC

Product Code. 30361536
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30361536 100 mg 100mg
30353010 10 mg 10mg
30359827 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30361536 Supplier Toronto Research Chemicals Supplier No. TRCC583535100MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25122-56-9
Molecular Formula C25 H30 Cl F O5
Molecular Weight (g/mol) 464.95
Synonym Clobetasone 17-Propionate, (16β)-21-Chloro-9-fluoro-16-methyl-3,11,20-trioxopregna-1,4-dien-17-yl Propionate, (16β)-21-Chloro-9-fluoro-16-methyl-17-(1-oxopropoxy)pregna-1,4-diene-3,11,20-trione, 21-Chloro-9-fluoro-17-hydroxy-16β-methylpregna-1,4-diene-3,11,20-trione propionate, Clobetasone propionate, Clobetasone-17-propionate
IUPAC Name [(8S,9R,10S,13S,14S,16S,17R)-17-(2-chloroacetyl)-9-fluoro-10,13,16-trimethyl-3,11-dioxo-7,8,12,14,15,16-hexahydro-6H-cyclopenta[a]phenanthren-17-yl] propanoate
SMILES CCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)C(=O)C[C@]12C)C(=O)CCl

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C25H30ClFO5/c1-5-21(31)32-25(20(30)13-26)14(2)10-18-17-7-6-15-11-16(28)8-9-22(15,3)24(17,27)19(29)12-23(18,25)4/h8-9,11,14,17-18H,5-7,10,12-13H2,1-4H3/t14-,17-,18-,22-,23-,24-,25-/m0/s1
Formula Weight 464.1766
Percent Purity >95%
Chemical Name or Material Clobetasone 17-Propionate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.