missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Clofazimine, 98%

Product Code. 15771779 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15771779 1 g 1g
15758817 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15771779 Supplier Thermo Scientific Chemicals Supplier No. 461760010
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2030-63-9 | C27H22Cl2N4 | 473.4 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2030-63-9
Molecular Formula C27H22Cl2N4
Molecular Weight (g/mol) 473.4
InChI Key WDQPAMHFFCXSNU-UHFFFAOYSA-N
PubChem CID 2794
ChEBI CHEBI:3749

Specifications

Melting Point 219°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in toluene,slightly soluble in ether,acetone,ethyl acetate
Formula Weight 473.4
Percent Purity 98%
Chemical Name or Material Clofazimine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.