missing translation for 'onlineSavingsMsg'
Learn More

Clofazimine, TRC

Product Code. 17920005
Change view
Click to view available options
Quantity:
50 mg
500 mg
1 g
Unit Size:
1g
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17920005 500 mg 500mg
17900095 50 mg 50mg
17909864 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17920005 Supplier Toronto Research Chemicals Supplier No. TRCC324300500MG
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2030-63-9
Molecular Formula C27 H22 Cl2 N4
Molecular Weight (g/mol) 473.4
Synonym Clofazimine, N,5-Bis(4-chlorophenyl)-3-[(1-methylethyl)imino]-3,5-dihydrophenazin-2-amine, 2-Phenazinamine, N,5-bis(4-chlorophenyl)-3,5-dihydro-3-[(1-methylethyl)imino]-, Phenazine, 3-(p-chloroanilino)-10-(p-chlorophenyl)-2,10-dihydro-2-(isopropylimino)- (6CI,7CI,8CI), N,5-Bis(4-chlorophenyl)-3,5-dihydro-3-[(1-methylethyl)imino]-2-phenazinamine, 2-(4-Chloroanilino)-3-isopropylimino-5-(4-chlorophenyl)-3,5-dihydrophenazine, 2-p-Chloroanilino-5-p-chlorophenyl-3,5-dihydro-3-isopropyliminophenazine, 3-(p-Chloroanilino)-10-(p-chlorophenyl)-2,10-dihydro-2-(isopropylimino)phenazine, B 663, B 663 (pharmaceutical), Chlofazimine, G 30320, Hansepran, Lampren, Lamprene, NSC 141046
IUPAC Name N,5-bis(4-chlorophenyl)-3-propan-2-yliminophenazin-2-amine
SMILES CC(C)N=C1C=C2N(c3ccc(Cl)cc3)c4ccccc4N=C2C=C1Nc5ccc(Cl)cc5

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C27H22Cl2N4/c1-17(2)30-24-16-27-25(15-23(24)31-20-11-7-18(28)8-12-20)32-22-5-3-4-6-26(22)33(27)21-13-9-19(29)10-14-21/h3-17,31H,1-2H3
Formula Weight 472.1222
Chemical Name or Material Clofazimine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.