missing translation for 'onlineSavingsMsg'
Learn More

Clofibrate, TRC

Product Code. 30364464
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30364464 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30364464 Supplier Toronto Research Chemicals Supplier No. TRCC5869101G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl clofibrate
CAS 637-07-0
Molecular Formula C12 H15 Cl O3
Quantity 1 g
Synonym Propionic acid, 2-(p-chlorophenoxy)-2-methyl-, ethyl ester (6CI,8CI), 2-(4-Chlorophenoxy)isobutyric acid ethyl ester, 2-(p-Chlorophenoxy)-2-methylpropionic acid ethyl ester, AY 61123, Abitrate, Amotril, Anparton, Apolan, Arteriosan, Artevil, Ateculon, Ateriosan, Atheropront, Atromid S, Atromidin, Azionyl, Bioscleran, CPIB, Cartagyl, Claripex, Claripex CPIB, Clobren SF, Clofibrat, Clofibrate, Clofinit, ECPIB, EPIB, Ethyl 2-(4-chlorophenoxy)-2-methylpropionate, Ethyl 2-(4-chlorophenoxy)isobutyrate, Ethyl 2-(p-chlorophenoxy)-2-methylpropionate, Ethyl 2-(p-chlorophenoxy)isobutyrate, Ethyl clofibrate, Ethyl p-chlorophenoxyisobutyrate, Ethyl α-(4-chlorophenoxy)-α-methylpropionate, Ethyl α-(4-chlorophenoxy)isobutyrate, Ethyl α-(p-chlorophenoxy)-α-methylpropionate, Ethyl α-(p-chlorophenoxy)isobutyrate, Hyclorate, ICI 28257, Lipavil, Lipavlon, Lipomid, Liprinal, Miscleron, Misclerone, NSC 79389, Neo-Atromid, Normet, Normolipol, Recolip, Regelan, Serotinex, Sklerepmexe, Sklero-Tablinene, Sklerolip, Skleromexe, Ticlobran, Xyduril, p-Chlorophenoxyisobutyric acid ethyl ester
InChI Formula InChI=1S/C12H15ClO3/c1-4-15-11(14)12(2,3)16-10-7-5-9(13)6-8-10/h5-8H,4H2,1-3H3
SMILES CCOC(=O)C(C)(C)Oc1ccc(Cl)cc1
IUPAC Name ethyl 2-(4-chlorophenoxy)-2-methylpropanoate
Molecular Weight (g/mol) 242.7
Formula Weight 242.071
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.