missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies ClonaCell™-HY Medium D

Product Code. 16447490
Encompass
Change view
Click to view available options
Quantity:
90 mL
Unit Size:
90mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16447490 90 mL 90mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16447490 Supplier STEMCELL Technologies Supplier No. 03804
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Semi-solid methylcellulose-based hybridoma selection and cloning medium with HAT (serum-containing)

  • Save research time and resources by performing hybridoma selection and cloning in one step with this methylcellulose-based medium
  • Use ClonaCell-HY Medium D to easily isolate individual clones with a high probability of monoclonality for further screening and expansion
  • Hybridoma clones typically have a wide range of growth rates and productivities; when cloned using ClonaCell-HY Medium D, they form visible and discrete monoclonal colonies, preventing the loss of rare and high-producing clones due to overgrowth
  • ClonaCell-HY Medium D is a semi-solid medium containing serum and the selective agents hypoxanthine, aminopterin, and thymidine (HAT)
  • It has been verified for use in developing mouse and rat hybridomas
  • It has also been reported to be compatible with producing and cloning hybridomas using lymphocytes from a variety of host animals, including human, mouse, rat, and hamster (e.g. Wilson JR et al. Antiviral Res, 2016)

Specifications

Cell Type Hybridomas
For Use With (Application) Cell Culture; Semi-Solid Cloning
Quantity 90 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.