missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies ClonaCell™-HY Medium E

Product Code. 17101036
Encompass
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17101036 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17101036 Supplier STEMCELL Technologies Supplier No. 03805
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hybridoma growth medium with hypoxanthine and thymidine (serum-containing)

  • ClonaCell™-HY Medium E is a serum-containing, nutritionally rich liquid medium optimized for hybridoma expansion after hypoxanthine, aminopterin, thymidine (HAT) selection
  • The medium contains hypoxanthine and thymidine (HT) and is used to wean hybridomas off aminopterin used during the selection process
  • This medium has been verified for use in mouse and rat hybridoma development and monoclonal antibody production and reportedly is compatible for production, cloning, and expansion of hybridomas using lymphocytes from a variety of host animals including human, mouse, rat, and hamster
  • ClonaCell™-HY Medium E has proven effective for subcloning and stabilization of unstable hybridoma cell lines

Specifications

Cell Type Hybridomas
For Use With (Application) Cell Culture; Hybridoma Generation
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.